Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione 97%

Supplier: AMBEED, INC
Warning

A122227-100MG A122227-1G A122227-250MG
76688-166EA 58 USD InStock
76688-166 76688-168 76688-170
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione 97%
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione
Precaución: for Research Use Only


Formula: C20H13N3O2
MW: 327.3361 g/mol
MDL Number: MFCD00236432
CAS Number: 119139-23-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty