Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nalpha-Acetyl-N-γ-trityl-L-asparagine 97%

Supplier: AMBEED, INC
Warning

A122499-100MG A122499-1G A122499-250MG
76688-172EA 5 USD InStock
76688-172 76688-174 76688-176
Nalpha-Acetyl-N-γ-trityl-L-asparagine 97%
Nalpha-Acetyl-N-gamma-trityl-L-asparagine
Precaución: for Research Use Only


Formula: C₂₅H₂₄N₂O₄
MDL Number: MFCD00236745
CAS Number: 163277-78-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty