Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-3,4-dehydro-L-proline 98%

Supplier: AMBEED, INC
Warning

A122651-100MG A122651-10G A122651-1G A122651-250MG A122651-5G
76688-182EA 9 USD InStock
76688-182 76688-184 76688-186 76688-188 76688-190
Boc-3,4-dehydro-L-proline 98%
Boc-3,4-dehydro-L-proline
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO₄
MDL Number: MFCD00037896
CAS Number: 51154-06-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty