Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-methoxyphenylacetate 98%

Supplier: AMBEED, INC
Warning

A124673-100G A124673-10G A124673-1G A124673-25G A124673-5G
76688-292EA 97 USD InStock
76688-292 76688-294 76688-296 76688-298 76688-300
Methyl-2-methoxyphenylacetate 98%
Methyl-2-methoxyphenylacetate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MDL Number: MFCD02093479
CAS Number: 27798-60-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty