Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Butoxyphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A125205-10G A125205-1G A125205-250MG A125205-25G A125205-5G
76688-352EA 49 USD InStock
76688-352 76688-354 76688-356 76688-358 76688-360
2-Butoxyphenylboronic acid 97%
2-Butoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₅BO₃
MW: 194.04 g/mol
MDL Number: MFCD03427052
CAS Number: 91129-69-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty