Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloromethyl butyrate 98%

Supplier: AMBEED, INC
Danger

A125683-25G A125683-5G
76688-384EA 19 USD InStock
76688-384 76688-386
Chloromethyl butyrate 98%
Chloromethyl butyrate
Precaución: for Research Use Only


Formula: C₅H₉ClO₂
MW: 136.58 g/mol
MDL Number: MFCD00216961
CAS Number: 33657-49-7
UN: 2924
ADR: 3,8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty