Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2-aminoacetoxy)copper 98%

Supplier: AMBEED, INC
Warning

A127549-100G A127549-10G A127549-500G A127549-25G
76688-502EA 12 USD InStock
76688-502 76688-504 76688-506 77313-224
Bis(2-aminoacetoxy)copper 98%
Bis(2-aminoacetoxy)copper
Precaución: for Research Use Only


Formula: C₄H₈CuN₂O₄
MW: 211.66 g/mol
MDL Number: MFCD00065114
CAS Number: 13479-54-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty