Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-Tyr(ME)-OH 97%

Supplier: AMBEED, INC
Warning

A127956-10G A127956-1G A127956-250MG A127956-25G A127956-5G
76688-520EA 53 USD InStock
76688-520 76688-522 76688-524 76688-526 76688-528
Boc-D-Tyr(ME)-OH 97%
Boc-D-Tyr(ME)-OH
Precaución: for Research Use Only


Formula: C₁₅H₂₁NO₅
MDL Number: MFCD00065604
CAS Number: 68856-96-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty