Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylcinnamic acid 98%

Supplier: AMBEED, INC
Warning

A128335-100G A128335-25G A128335-500G A128335-5G
76688-538EA 132 USD InStock
76688-538 76688-540 76688-542 76688-544
3-Methylcinnamic acid 98%
3-Methylcinnamic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MDL Number: MFCD00016844
CAS Number: 3029-79-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty