Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-7-benzyl-9-oxo-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate 95%

Supplier: AMBEED, INC
Warning

A132358-1G A132358-250MG A132358-5G
76688-682EA 64 USD InStock
76688-682 76688-684 76688-686
tert-Butyl-7-benzyl-9-oxo-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate 95%
tert-Butyl-7-benzyl-9-oxo-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₉H₂₆N₂O₃
MDL Number: MFCD19443919
CAS Number: 227940-70-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty