Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Aminoethyl)-2-(4-(2-(4-((9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-yl)amino)phenyl)acetamido)phenyl)acetamide 98+%

Supplier: AMBEED, INC
Warning

A132999-10MG A132999-1MG A132999-50MG A132999-5MG
76688-708EA 109 USD InStock
76688-708 76688-710 76688-712 76688-714
N-(2-Aminoethyl)-2-(4-(2-(4-((9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-yl)amino)phenyl)acetamido)phenyl)acetamide 98+%
N-(2-Aminoethyl)-2-(4-(2-(4-((9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-yl)amino)phenyl)acetamido)phenyl)acetamide
Precaución: for Research Use Only


Formula: C₂₈H₃₂N₈O₆
MDL Number: MFCD00055047
CAS Number: 96760-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty