Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Butoxyaniline 97%

Supplier: AMBEED, INC
Warning

A135979-10G A135979-1G A135979-25G
76688-808EA 33 USD InStock
76688-808 76688-810 76688-812
4-Butoxyaniline 97%
4-Butoxyaniline
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO
MDL Number: MFCD00007866
CAS Number: 4344-55-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty