Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

19-Bromononadecanoic acid 97%

Supplier: AMBEED, INC
Warning

A136235-1G A136235-250MG
76688-822EA 225 USD InStock
76688-822 76688-824
19-Bromononadecanoic acid 97%
19-Bromononadecanoic acid
Precaución: for Research Use Only


Formula: C₁₉H₃₇BrO₂
MDL Number: MFCD00082606
CAS Number: 78774-38-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty