Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3,4-difluorobenzoate 98%

Supplier: AMBEED, INC
Warning

A139275-100G A139275-10G A139275-25G A139275-500G A139275-5G
76688-956EA 91 USD InStock
76688-956 76688-958 76688-960 76688-962 76688-964
Ethyl-3,4-difluorobenzoate 98%
Ethyl-3,4-difluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₈F₂O₂
MDL Number: MFCD00272138
CAS Number: 144267-96-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty