Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-5-methylbenzamide 95%

Supplier: AMBEED, INC
Warning

A140736-10G A140736-1G A140736-5G
76689-004EA 129 USD InStock
76689-004 76689-006 76689-008
2-Hydroxy-5-methylbenzamide 95%
2-Hydroxy-5-methylbenzamide
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00017124
CAS Number: 39506-61-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty