Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Quinolinediol 98%

Supplier: AMBEED, INC
Warning

A141693-100G A141693-10G A141693-1G A141693-25G A141693-5G
76689-022EA 171 USD InStock
76689-022 76689-024 76689-026 76689-028 76689-030
2,6-Quinolinediol 98%
2,6-Quinolinediol
Precaución: for Research Use Only


Formula: C₉H₇NO₂
MDL Number: MFCD00239446
CAS Number: 19315-93-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty