Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyltributylammonium bromide 98%

Supplier: AMBEED, INC
Warning

A148994-100G A148994-500G
76689-286EA 55 USD InStock
76689-286 76689-288 76689-290
Benzyltributylammonium bromide 98%
Benzyltributylammonium bromide
Precaución: for Research Use Only


Formula: C₁₉H₃₄BrN
MDL Number: MFCD00011848
CAS Number: 25316-59-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty