Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-2-(trifluoromethyl)benzaldehyde 98%

Supplier: AMBEED, INC
Warning

A149267-10G A149267-1G A149267-250MG A149267-5G
76689-306EA 47 USD InStock
76689-306 76689-308 76689-310 76689-312
4-Methoxy-2-(trifluoromethyl)benzaldehyde 98%
4-Methoxy-2-(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only


Formula: C₉H₇F₃O₂
MDL Number: MFCD01091011
CAS Number: 106312-36-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty