Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium (3S,4R,5R)-6-((hydrogenphosphonato)oxy)-3,4,5-trihydroxy-2-oxohexyl phosphate xhydrate 97%

Supplier: AMBEED, INC
Warning

A151466-10G A151466-1G A151466-5G
76689-364EA 15 USD InStock
76689-364 76689-366 76689-368
Sodium (3S,4R,5R)-6-((hydrogenphosphonato)oxy)-3,4,5-trihydroxy-2-oxohexyl phosphate xhydrate 97%
Sodium (3S,4R,5R)-6-((hydrogenphosphonato)oxy)-3,4,5-trihydroxy-2-oxohexyl phosphate xhydrate
Precaución: for Research Use Only


Formula: C₆H₁₃Na₃O₁₃P₂
MDL Number: MFCD00068321
CAS Number: 41012-14-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty