Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Avasimibe 95%

Supplier: AMBEED, INC
Warning

A156472-100MG A156472-10MG A156472-1MG A156472-250MG A156472-50MG A156472-5MG
76689-568EA 460 USD InStock
76689-568 76689-570 76689-572 76689-574 76689-576 76689-578
Avasimibe 95%
Avasimibe
Precaución: for Research Use Only


Formula: C₂₉H₄₃NO₄S
MDL Number: MFCD00934956
CAS Number: 166518-60-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty