Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylpentyl benzoate 97%

Supplier: AMBEED, INC
Warning

A157107-250MG A157107-5G
76689-604EA 23 USD InStock
76689-604 76689-606
2-Methylpentyl benzoate 97%
2-Methylpentyl benzoate
Precaución: for Research Use Only


Formula: C₁₃H₁₈O₂
MW: 206.28 g/mol
MDL Number: MFCD00059413
CAS Number: 59736-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty