Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Clemastine fumarate 98%

Supplier: AMBEED, INC
Warning

A157173-100MG A157173-1MG A157173-250MG A157173-25MG A157173-1G
76689-610EA 29 USD InStock
76689-610 76689-612 76689-614 76689-616 77625-462
Clemastine fumarate 98%
Clemastine fumarate
Precaución: for Research Use Only


Formula: C₂₅H₃₀ClNO₅
MW: 459.9600 g/mol
MDL Number: MFCD00137486
CAS Number: 14976-57-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty