Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-allyl-Gly-OH dicyclohexylammonium salt 97%

Supplier: AMBEED, INC
Warning

A158024-1G A158024-5G
76689-664EA 95.49 USD InStock
76689-664 76689-666 76689-668
Boc-D-allyl-Gly-OH dicyclohexylammonium salt 97%
Boc-D-allyl-Gly-OH dicyclohexylammonium salt
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₄·C₁₂H₂₃N
MDL Number: MFCD01321012
CAS Number: 221352-64-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty