Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenylcyclopropanecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A159014-100G A159014-10G A159014-1G A159014-250MG A159014-25G A159014-5G
76689-736EA 121 USD InStock
76689-736 76689-738 76689-740 76689-742 76689-744 76689-746
1-Phenylcyclopropanecarboxylic acid 98%
1-Phenylcyclopropanecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD00001288
CAS Number: 6120-95-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty