Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,8-Quinolinediol 98%

Supplier: AMBEED, INC
Warning

A164072-100G A164072-10G A164072-1G A164072-25G A164072-500G A164072-5G
76689-932EA 87 USD InStock
76689-932 76689-934 76689-936 76689-938 76689-940 76689-942
2,8-Quinolinediol 98%
2,8-Quinolinediol
Precaución: for Research Use Only


Formula: C₉H₇NO₂
MW: 161.16
MDL Number: MFCD00216696
CAS Number: 15450-76-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty