Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Homophenylalanine 98%

Supplier: AMBEED, INC
Warning

A165377-10G A165377-25G A165377-5G A165377-100G
76690-014EA 24 USD InStock
76690-014 76690-016 76690-018 77625-628
DL-Homophenylalanine 98%
DL-Homophenylalanine
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD00063092
CAS Number: 1012-05-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty