Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl isovalerate 98%

Supplier: AMBEED, INC
Warning

A173423-1000G A173423-100G A173423-25G A173423-500G
76690-242EA 115 USD InStock
76690-242 76690-244 76690-246 76690-248
Benzyl isovalerate 98%
Benzyl isovalerate
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
MDL Number: MFCD00048348
CAS Number: 103-38-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty