Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Creatinine 98%

Supplier: AMBEED, INC
Warning

A174549-100G A174549-10G A174549-25G A174549-500G A174549-1000G
76690-270EA 44 USD InStock
76690-270 76690-272 76690-274 76690-276 76690-278 77625-826
Creatinine 98%
Creatinine
Precaución: for Research Use Only


Formula: C₄H₇N₃O
MW: 113.12 g/mol
MDL Number: MFCD00059730
CAS Number: 60-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty