Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-((4-(Mesitylamino)pyrimidin-2-yl)amino)benzonitrile 98%

Supplier: AMBEED, INC
Warning

A175194-10MG A175194-1MG A175194-50MG A175194-5MG
76690-310EA 18 USD InStock
76690-310 76690-312 76690-314 76690-316
4-((4-(Mesitylamino)pyrimidin-2-yl)amino)benzonitrile 98%
4-((4-(Mesitylamino)pyrimidin-2-yl)amino)benzonitrile
Precaución: for Research Use Only


Formula: C₂₀H₁₉N₅
MDL Number: MFCD09833899
CAS Number: 244767-67-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty