Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9,10-Bis(diethylphosphonomethyl)anthracene 95%

Supplier: AMBEED, INC
Warning

A175650-100MG A175650-1G A175650-250MG A175650-5G
76690-344EA 8 USD InStock
76690-344 76690-346 76690-348 76690-350
9,10-Bis(diethylphosphonomethyl)anthracene 95%
9,10-Bis(diethylphosphonomethyl)anthracene
Precaución: for Research Use Only


Formula: C₂₄H₃₂O₆P₂
MDL Number: MFCD08276315
CAS Number: 60974-92-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty