Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Isoquinolineboronic acid 97%

Supplier: AMBEED, INC
Warning

A176622-100MG A176622-1G A176622-250MG A176622-5G
76690-368EA 29 USD InStock
76690-368 76690-370 76690-372 76690-374
8-Isoquinolineboronic acid 97%
8-Isoquinolineboronic acid
Precaución: for Research Use Only


Formula: C₉H₈BNO₂
MW: 172.9800 g/mol
MDL Number: MFCD11044913
CAS Number: 721401-43-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty