Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-3-(4-methylphenyl)propionic acid 97%

Supplier: AMBEED, INC
Warning

A177837-10G A177837-25G A177837-5G
76690-434EA 37 USD InStock
76690-434 76690-436 76690-438 77625-834
3-Amino-3-(4-methylphenyl)propionic acid 97%
3-Amino-3-(4-methylphenyl)propionic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₂
MW: 179.2157 g/mol
MDL Number: MFCD01863249
CAS Number: 68208-18-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty