Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1E,6E)-1,7-Bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione 98%

Supplier: AMBEED, INC
Warning

A178092-10MG A178092-1G A178092-1MG A178092-250MG A178092-25MG A178092-50MG A178092-5G
76690-450EA 8 USD InStock
76690-450 76690-452 76690-454 76690-456 76690-458 76690-460 76690-462
(1E,6E)-1,7-Bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione 98%
(1E,6E)-1,7-Bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione
Precaución: for Research Use Only


Formula: C₁₉H₁₆O₄
MDL Number: MFCD03419284
CAS Number: 33171-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty