Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methylamino)-1-phenylpropan-1-one hydrochloride 97%

Supplier: AMBEED, INC
Warning

A178544-100MG A178544-1G A178544-250MG
76690-474EA 26 USD InStock
76690-474 76690-476 76690-478
3-(Methylamino)-1-phenylpropan-1-one hydrochloride 97%
3-(Methylamino)-1-phenylpropan-1-one hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₄ClNO
MW: 199.68 g/mol
MDL Number: MFCD08063896
CAS Number: 2538-50-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty