Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl acetonylsuccinate 97%

Supplier: AMBEED, INC
Warning

A180630-10G A180630-1G A180630-25G A180630-5G
76690-544EA 43 USD InStock
76690-544 76690-546 76690-548 76690-550
Diethyl acetonylsuccinate 97%
Diethyl acetonylsuccinate
Precaución: for Research Use Only


Formula: C₁₁H₁₈O₅
MW: 230.26 g/mol
MDL Number: MFCD00071579
CAS Number: 1187-74-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty