Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

β-Nicotinamide adenine dinucleotide phosphate (β-NADP) 98%

Supplier: AMBEED, INC
Warning

A181468-1G A181468-25G A181468-5G A181468-100G
76690-576EA 27 USD InStock
76690-576 76690-578 76690-580 77625-864
β-Nicotinamide adenine dinucleotide phosphate (β-NADP) 98%
ß-Nicotinamide adenine dinucleotide phosphate (β-NADP)
Precaución: for Research Use Only


Formula: C₂₁H₂₈N₇O₁₇P₃
MDL Number: MFCD00067537
CAS Number: 53-59-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty