Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(4-hydroxyphenyl) disulfide 98%

Supplier: AMBEED, INC
Warning

A183502-100G A183502-1G A183502-25G A183502-500G A183502-5G
76690-638EA 201 USD InStock
76690-638 76690-640 76690-642 76690-644 76690-646
Bis(4-hydroxyphenyl) disulfide 98%
Bis(4-hydroxyphenyl)disulphide
Precaución: for Research Use Only


Formula: C₁₂H₁₀O₂S₂
MDL Number: MFCD00020164
CAS Number: 15015-57-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty