Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopropyldiphenylsulfonium tetrafluoroborate 95%

Supplier: AMBEED, INC
Warning

A184873-10G A184873-250MG A184873-5G
76690-682EA 396 USD InStock
76690-682 76690-684 76690-686
Cyclopropyldiphenylsulfonium tetrafluoroborate 95%
Cyclopropyldiphenylsulfonium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₁₅H₁₅BF₄S
MDL Number: MFCD00011873
CAS Number: 33462-81-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty