Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzoyl-DL-valine 95%

Supplier: AMBEED, INC
Warning

A185053-10G A185053-1G A185053-25G A185053-5G
76690-688EA 11 USD InStock
76690-688 76690-690 76690-692 76690-694
Benzoyl-DL-valine 95%
Benzoyl-DL-valine
Precaución: for Research Use Only


Formula: C₁₂H₁₅NO₃
MW: 221.26 g/mol
MDL Number: MFCD00038283
CAS Number: 2901-80-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty