Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitro-m-anisic acid 98%

Supplier: AMBEED, INC
Warning

A188777-100G A188777-10G A188777-25G A188777-5G A188777-1G
76690-784EA 48 USD InStock
76690-784 76690-786 76690-788 76690-790 77625-902
4-Nitro-m-anisic acid 98%
4-Nitro-m-anisic acid
Precaución: for Research Use Only


Formula: C₈H₇NO₅
MW: 197.15 g/mol
Boiling Pt: 405
Melting Pt: 233
MDL Number: MFCD00007353
CAS Number: 5081-36-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty