Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Amino-1-tetralone 97%

Supplier: AMBEED, INC
Warning

A188850-100G A188850-10G A188850-1G A188850-250MG A188850-25G A188850-5G
76690-792EA 239 USD InStock
76690-792 76690-794 76690-796 76690-798 76690-800 76690-802
6-Amino-1-tetralone 97%
6-Amino-1-tetralone
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
MDL Number: MFCD00099462
CAS Number: 3470-53-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty