Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Baclofen 98%

Supplier: AMBEED, INC
Danger

A190802-100G A190802-10G A190802-1G A190802-1MG A190802-25G A190802-5G
76690-876EA 231 USD InStock
76690-876 76690-878 76690-880 76690-882 76690-884 76690-886
Baclofen 98%
Baclofen
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClNO₂
MDL Number: MFCD00055143
CAS Number: 1134-47-0
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty