Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminophenylboronic acid hemisulfate 97%

Supplier: AMBEED, INC
Warning

A191297-10G A191297-1G A191297-25G A191297-5G
76690-906EA 30 USD InStock
76690-906 76690-908 76690-910 76690-912
3-Aminophenylboronic acid hemisulfate 97%
3-Aminophenylboronic acid hemisulphate
Precaución: for Research Use Only


Formula: C₁₂H₁₈B₂N₂O₈S
MDL Number: MFCD00013111
CAS Number: 66472-86-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty