Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodo-2,6-xylenol 97%

Supplier: AMBEED, INC
Warning

A192064-100G A192064-10G A192064-1G A192064-25G A192064-5G
76690-914EA 73 USD InStock
76690-914 76690-916 76690-918 76690-920 76690-922
4-Iodo-2,6-xylenol 97%
4-Iodo-2,6-xylenol
Precaución: for Research Use Only


Formula: C₈H₉IO
MDL Number: MFCD00094415
CAS Number: 10570-67-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty