Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-3-Phenylprop-2-en-1-ol 97%

Supplier: AMBEED, INC
Warning

A194226-100G A194226-25G A194226-5G A194226-500G A194226-1000G
76690-982EA 10 USD InStock
76690-982 76690-984 76690-986 77567-814 77625-950
(E)-3-Phenylprop-2-en-1-ol 97%
(E)-3-Phenylprop-2-en-1-ol
Precaución: For research use only.


Formula: C₉H₁₀O
MDL Number: MFCD00002921
CAS Number: 4407-36-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty