Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl cyclopentylmalonate 95%

Supplier: AMBEED, INC

A199614-1G A199614-25G A199614-5G
76691-120EA 14 USD InStock
76691-120 76691-122 76691-124
Dimethyl cyclopentylmalonate 95%
Dimethyl cyclopentylmalonate
Precaución: for Research Use Only


Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
MDL Number: MFCD03844781
CAS Number: 82491-60-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty