Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-2,3-dimethylbenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A200012-10G A200012-1G A200012-250MG A200012-25G A200012-5G
76691-128EA 62 USD InStock
76691-128 76691-130 76691-132 76691-134 76691-136
4-Methoxy-2,3-dimethylbenzaldehyde 97%
4-Methoxy-2,3-dimethylbenzaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
MDL Number: MFCD00003339
CAS Number: 38998-17-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty