Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Caronic anhydride 95%

Supplier: AMBEED, INC
Warning

A201024-100G A201024-10G A201024-1G A201024-25G A201024-5G
76691-178EA 49 USD InStock
76691-178 76691-180 76691-182 76691-184 76691-186
Caronic anhydride 95%
Caronic anhydride
Precaución: for Research Use Only


Formula: C₇H₈O₃
MW: 140.14 g/mol
MDL Number: MFCD09751198
CAS Number: 67911-21-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty