Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II) 98%

Supplier: AMBEED, INC
Warning

A202094-100MG A202094-1G A202094-250MG A202094-5G
76691-228EA 5 USD InStock
76691-228 76691-230 76691-232 76691-234
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II) 98%
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)
Precaución: for Research Use Only


Formula: C₅₅H₄₇OP₃Ru
MDL Number: MFCD00015870
CAS Number: 25360-32-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty