Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-(chloromethyl)pyridine 95%

Supplier: AMBEED, INC
Danger

A204006-100MG A204006-1G A204006-250MG
76691-302EA 27 USD InStock
76691-302 76691-304 77626-036
2-Chloro-6-(chloromethyl)pyridine 95%
2-Chloro-6-(chloromethyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
MDL Number: MFCD07774109
CAS Number: 78846-88-5
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty